missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 986187, >98%, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent δ-opioid receptor positive allosteric modulator (EC50 = 30 nM). Exhibits 100-fold selectivity for δ over μ-opioid receptor. Enhances affinity of [Leu5]-Enkephalin (Cat.No. 1889), SNC 80 (Cat.No. 0764) and TAN 67 (Cat.No. 0921) for the δ-opioid receptor.
Specifications
Linear Formula | C31H34O4 |
Quantity | 10 mg |
Solubility Information | Soluble to 20mM in DMSO |
Formula Weight | 470.61 |
Chemical Name or Material | 3,4,5,6,7,9-Hexahydro-3,3,6,6-tetramethyl-9-[4-[(2-methylphenyl)methoxy]phenyl]-1H-xanthene-1,8(2H)- |
Grade | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'