missing translation for 'onlineSavingsMsg'
Learn More

IKA™ BMT-20-S-M-Gamma Tube for Grinding with Stainless Steel Balls (20x)

For milling of dry, brittle samples, cell maceration, processing of materials mixed with fluids.

Brand:  IKA™ 0003700701

Additional Details : Weight : 0.72000kg

Product Code. 15812588

  • £267.00 / Each
Estimated Shipment: 30-09-2024
to see stock.
Add to basket

Description

Description

  • Components: Tube, stainless steel balls, cover
  • Application: Dry milling of dry and brittle samples (e.g., kaolin, gypsum, colored pigments, tablets), cell maceration, processing of materials mixed with fluids
  • Further suitable for:
    • additional media quantities may be added during operation
    • sampling during operation
    • sample preparation under sterile conditions
    • germ free conservation of samples (tissue, blood, etc.)
Specifications

Specifications

2 to 15 mL
Tube, Stainless Steel balls, cover
20/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers