Learn More
IKA™ BMT-20-S Tubes
Brand: IKA™ 0003703200
Code : 50
Additional Details : Weight : 1.00000kg
Specifications
BMT-20-S Tube | |
Dry milling of dry and brittle samples (e.g. kaolin, gypsum, colored pigments, tablets), cell maceration, processing of materials mixed with fluids | |
25 Pk. |
2 to 15 mL | |
BMT-20-S |