missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ BNC to BNC Connection Cable

25 ft BNC-BNC connection/extension cable for ideally for connecting pH electrodes.

Brand:  Fisherbrand™ 99001048-025B

Product Code. 11739539


Description

Description

  • 25 ft BNC extension/connection cable
  • BNC-BNC plug and adapter converter
  • Length 7.6 m

Specifications

Specifications

BNC to BNC Connection Cable
7.6 m
7.6 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers