missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Boc-D(OMe)-FMK Inhibitor
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Specifications
Specifications
| Color | White |
| For Use With (Application) | Blocking/Neutralizing, In vitro assay, In vivo assay |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles |
| Quantity | 5 mg |
| Product Type | Boc-D(OMe)-FMK Inhibitor |
| Molecular Weight (g/mol) | 263 |
| Inhibitors | Boc-D(OMe)-FMK |
| Form | Semi-solid |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction