missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-Proline, 99+%, Thermo Scientific Chemicals

Product Code. 10016153
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15761-39-4
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00037324
InChI Key ZQEBQGAAWMOMAI-ZETCQYMHSA-N
Synonym boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro
PubChem CID 85083
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 133.0°C to 136.0°C
Color White
Assay Percent Range 99+%
Infrared Spectrum Authentic
Beilstein 22, 35
Packaging Glass Bottle
Specific Rotation − 60.50
Formula Weight 215.25
Percent Purity ≥99%
Water 0.5% max. (K.F.)
Specific Rotation Condition −60.5° (20°C 589nm,c=2,CH3COOH)
Quantity 100 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material BOC-L-Proline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'