missing translation for 'onlineSavingsMsg'
Learn More
BOC-L-Tyrosine, 99+%, Thermo Scientific Chemicals Product Code.: 10571491

BOC-L-Tyrosine, 99+%, Thermo Scientific Chemicals

Product Code. 10571491
25 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10571491

Brand: Thermo Scientific Acros 275700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3978-80-1
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.31
MDL Number MFCD00037179
InChI Key CNBUSIJNWNXLQQ-LDGXTIHJNA-N
Synonym boc-tyr-oh, boc-l-tyrosine, n-boc-l-tyrosine, n-tert-butoxycarbonyl-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl, chembl65386, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosineShow MoreShow Less
PubChem CID 117439
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 135.0°C to 140.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula HOC6H4CH2CH(COOH)NHCO2C(CH3)3
Specific Rotation + 2.50
Water 1% Max. (K.F.)
IUPAC Name (2S)-3-(4-hydroxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
Quantity 25 g
Formula Weight 281.3
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material BOC-L-Tyrosine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461381 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number275701000,275700250,275700050
Lot NumberA0459508 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number275701000,275700250,275700050
Lot NumberA0384275 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number275701000,275700250,275700050
Lot NumberA0411483 Certificate TypeCertificate of Analysis Date21/02/2022 Catalog Number275701000,275700250,275700050
Lot NumberA0423799 Certificate TypeCertificate of Analysis Date28/10/2020 Catalog Number
Product Identifier
  • BOC-L-Tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BOC-L-Tyrosine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.