missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOC-O-Benzyl-L-threonine hydroxysuccinimide ester, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 104°C to 107°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 5 g |
Specific Rotation | 10.6 |
Formula Weight | 406.44 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | BOC-O-Benzyl-L-threonine hydroxysuccinimide ester |
Safety and Handling
Product Identifier
- BOC-O-Benzyl-L-threonine hydroxysuccinimide ester
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'