Learn More
BOC-ON, 99%, Thermo Scientific Chemicals
£30.10 - £318.00
Chemical Identifiers
CAS | 58632-95-4 |
---|---|
Molecular Formula | C13H14N2O3 |
Molecular Weight (g/mol) | 246.27 |
MDL Number | MFCD00001863 |
InChI Key | QQWYQAQQADNEIC-PTNGSMBKSA-N |
Synonym | 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino |
PubChem CID | 5868400 |
IUPAC Name | tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate |
SMILES | CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10550014
|
Thermo Scientific Acros
185890050 |
5 g | Glass bottle |
£30.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10591821
|
Thermo Scientific Acros
185890250 |
25 g | Glass bottle |
£91.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10594644
|
Thermo Scientific Acros
185891000 |
100 g | Glass bottle |
£318.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
58632-95-4 | |
246.27 | |
QQWYQAQQADNEIC-PTNGSMBKSA-N | |
5868400 | |
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1 |
C13H14N2O3 | |
MFCD00001863 | |
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino | |
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate |
Specifications
58632-95-4 | |
White to Beige | |
Authentic | |
C13H14N2O3 | |
MFCD00001863 | |
15, 1323 | |
Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,ether,benzene,dioxane,,chloroform,acetone,methanol,isopropanol and,tert-butanol,insoluble in petroleum ether | |
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1 | |
246.27 | |
246.27 | |
Fine Crystalline Powder |
86°C to 89°C | |
>100°C | |
99% | |
(CH3)3COCO2N=C(C6H5)CN | |
06,91; 10,61 | |
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino | |
QQWYQAQQADNEIC-PTNGSMBKSA-N | |
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate | |
5868400 | |
99% | |
BOC-ON, 99% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapo
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 261-370-9
missing translation for 'tsca' : TSCA
- BOC-ON
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only