Learn More
Bohlender™ BOLA™ Conductors for Magnetic Stirrer Heads
Conductors for Magnetic Stirrer Heads made of borosilicate glass.
Brand:  Bohlender™ C463-45
 View more versions of this product
Product Code. 17167134
  • £1190.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass with ground joint and glued upper bearing
  • Alternatively without ground joint for fusing on the reactor lid (upper bearing to be glued is included).
Specifications
Specifications
NS 45/40
NS 45/40
Borosilicate Glass
Left-handed Thread M 21 x 1.75