Learn More
Bohlender™ BOLA™ Lid for Flat Flange Vessels
Lid for Flat Flange Vessels made of PTFE with center neck socket NS 29/32 and three lateral necks.
Brand: Bohlender™ B326-01
This item is not returnable.
View return policy
Description
- Suitable for reactor made of PFA (Cat. No. B 325-01)
- With socket center neck NS 29/32 and three lateral necks with ground joint sockets NS 29/32
- Bottom side with center collar for non-slipping alignment on the reactor
- All lateral connectors are made for an insertion of tubes and probes aside the centre in order to avoid collisions
Specifications
PTFE | |
Flat Flange NW 170/146 | |
Lateral Necks NS 3 x 29/32 Aside the Shaft |