missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PTFE Receiver Adapter

Designed with an integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint. BOLA PTFE Receiver Adapeters feature universal chemical resistance.

Brand:  Bohlender™ B292-02

Product Code. 10013321

  • £183.00 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PTFE Reciever Adaptor
  • Ground joint cone and socket size 29
  • Additional socket size 19 for vacuum stopcock (BOLA Cat. No. B 293-02)

Compliance

FDA

Specifications

Specifications

FDA
Cone: NS 29/32; Socket: NS 19/26
Leteral Socket: NS 29/32
250°C
Receiver Adapter
-200°C
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bohlender™ BOLA™ PTFE Receiver Adapter > PTFE Receiver Adapters

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.