Learn More
Bohlender™ BOLA™ Sleeves with Gripping Ring
Click to view available options
Standard Taper Joint No.:
US 24/40
Unit Size:
Pack of 2
Description
- Made of PTFE
- Gripping ring and sealing rings on the outside
- High-vacuum tight sealing
- Low leak rate: <1 x 0.0001 mbar x l x s-1
- Solid construction for continued use
- Sealing rings are easy accessible and therefore easy to clean
Specifications
Specifications
| Material | PTFE |
| Standard Taper Joint No. | US 24/40 |
| Type | Sleeve with Gripping Ring |