Learn More
Bohlender™ BOLA™ Sleeves with Ribs
Product Code. 17117394
Click to view available options
Standard Taper Joint No.:
NS 10/19
NS 12/21
NS 24/29
NS 34/35
NS 40/38
US 14/20
US 24/40
Unit Size:
Pack of 10
Pack of 5
Product Code. 17117394
Brand: Bohlender™ H93006
Only null left
Add to basket
Add to basket
Sleeves with ribs for vacuum up to 0.1 mbar.
  • Sealing rings ('ribs') on the outside
  • Without gripping ring and therefore usable with joint clamps
  • For vacuum up to 0.1 mbar
  • Solid construction for continued use
  • Sealing rings are easily accessible and easy to clean
Specifications
For Use With (Equipment) Vacuum Up to 0.1 mbar
Material PTFE
Product Type Sleeves with Ribs
Standard Taper Joint No. NS 24/29