Learn More
Bohlender™ BOLA™ Sleeves with Ribs
Click to view available options
Standard Taper Joint No.:
NS 10/19
NS 12/21
NS 24/29
NS 34/35
NS 40/38
US 14/20
US 24/40
Unit Size:
Pack of 10
Pack of 5
Description
- Sealing rings ('ribs') on the outside
- Without gripping ring and therefore usable with joint clamps
- For vacuum up to 0.1 mbar
- Solid construction for continued use
- Sealing rings are easily accessible and easy to clean
Specifications
Specifications
| For Use With (Equipment) | Vacuum Up to 0.1 mbar |
| Material | PTFE |
| Product Type | Sleeves with Ribs |
| Standard Taper Joint No. | US 14/20 |