Learn More
Bohlender™ BOLA™ Spare O-rings for Stopcocks
Spare o-rings for stopcocks.
Brand: Bohlender™ J1198-01
This item is not returnable.
View return policy
Description
- O-rings made of NBR
- For BOLA™ Eco Stopcock Plugs
- Taper: 1:10
- Thread M: 7 mm
- Suitable for size: NS 10
Specifications
Spare O-ring | |
NBR | |
For BOLA™ Eco Stopcock Plugs |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Bohlender™ BOLA™ Spare O-rings for Stopcocks > Spare o-rings for stopcocks; Material: NBR; For Use With (Equipment): For BOLA Eco Stopcock Plugs; Type: Spare O-rings
Spot an opportunity for improvement?Share a Content Correction