Learn More
Bohlender™ BOLA™ Spherical Ground Joint Sleeves
Product Code. 17137404
Click to view available options
Standard Taper Joint No.:
S 13
S 19
S 29
S 40
Unit Size:
Pack of 2
Pack of 3
Product Code. 17137404
Brand: Bohlender™ H93402
Only null left
Add to basket
Add to basket
Spherical ground joint sleeves with gripping ring and sealing rings on the outside.
  • With gripping ring and sealing rings on the outside
  • High-vacuum tight sealing
  • Low leak rate: <1 x 0.01 mbar x l x s-1
  • Solid construction for continued use
Specifications
Material PTFE
Product Type Ground Joint Sleeves
Standard Taper Joint No. S 13