missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Bologna Brogue-Style Business Shoes
With ankle support in the shank.
£158.00 - £161.00
Specifications
Shank Type | Ankle support |
---|---|
Sole Features | Anti-bacterial with leather top layer inlay sole |
Sole Material | Impenetrable nitrile rubber |
Style | Brogue |
Certifications/Compliance (Europe) | EN ISO 20345:2011; ESD 0,1-35 M Ohm/NEN-EN-IEC 61340-5-1 |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
10565384
|
Emma Safety Footwear
MM10709048 |
48 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10526944
|
Emma Safety Footwear
MM10709043 |
43 |
£161.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10133944
|
Emma Safety Footwear
MM10709041 |
41 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10174204
|
Emma Safety Footwear
MM10709046 |
46 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10769425
|
Emma Safety Footwear
MM10709044 |
44 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10226084
|
Emma Safety Footwear
MM10709040 |
40 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10759425
|
Emma Safety Footwear
MM10709042 |
42 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10547114
|
Emma Safety Footwear
MM10709045 |
45 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10021944
|
Emma Safety Footwear
MM10709047 |
47 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
10779255
|
Emma Safety Footwear
MM10709039 |
39 |
£158.00
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||||
Description
- Polished black full-grain leather uppers
- Impenetrable nitrile rubber sole
- Hydro-Tec™ sanitized silver and wellness leather inlay sole
- Woven zero penetration insole
- Anti-bacterial with leather top layer in-lay sole
- Antistatic properties: ESD 0,1 - 35 M Ohm / NEN-EN-IEC 61340-5-1
Specifications
Ankle support | |
Impenetrable nitrile rubber | |
EN ISO 20345:2011; ESD 0,1-35 M Ohm/NEN-EN-IEC 61340-5-1 |
Anti-bacterial with leather top layer inlay sole | |
Brogue | |
Business Shoe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Emma Safety Footwear Bologna Brogue-Style Business Shoes