Learn More
Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications.
£133.00
Specifications
For Use With (Equipment) | Mini Gel Tank, XCell SureLock Mini-Cell |
---|---|
Gel Percentage | 10% |
Gel Size | Mini |
GelThickness | 1.0 mm |
Gel Type | Bis-Tris Plus |
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wells | Price | Quantity & Availability | |||||
15324594
|
Invitrogen™
NW00100BOX |
10-well |
£133.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15334594
|
Invitrogen™
NW00102BOX |
12-well |
£133.00
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15344594
|
Invitrogen™
NW00105BOX |
15-well |
£133.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
15354594
|
Invitrogen™
NW00107BOX |
17-well |
£133.00
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt Bis-Tris Plus gels to obtain optimal results for day-to-day protein separation needs.
Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES or MOPS SDS running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.
• Optimized run conditions—separate proteins using constant voltage in approximately 20 minutes
• Better protein integrity—neutral pH sample preparation process preserves proteins
• Improved protein separation—offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than tris-glycine based gels
• Wedge-shaped wells—easier sample loading and up to two times higher sample volume than other 1.0mm-thick gels
Specifications
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Bis-Tris Plus | |
3.5 to 260 kDa | |
Molecular Weight | |
10 Gels/Box | |
Store at 4–25°C. Do not freeze. | |
8 cm |
10% | |
1.0 mm | |
Denaturing | |
Denaturing | |
Bolt™, WedgeWell™ | |
Room Temperature | |
8 cm |