missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format

Invitrogen Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions.

£141.00

Specifications

For Use With (Equipment) Mini Gel Tank, XCell SureLock Mini-Cell
Gel Percentage 8%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris Plus
View More Specs

Requires: Mini Gel Tank (Mfr. No. A25977)


Products 4
Product Code Brand Wells Price Quantity & Availability  
Product Code Brand Wells Price Quantity & Availability  
15349473
View Documents Promotion Details
Invitrogen™
NW00080BOX
10-well
£141.00
Pack of 10
In Stock
to see stock.
Add to basket
 
15394584
View Documents Promotion Details
Invitrogen™
NW00082BOX
12-well
£141.00
Pack of 10
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
15304594
View Documents Promotion Details
Invitrogen™
NW00085BOX
15-well
£141.00
Pack of 10
In Stock
to see stock.
Add to basket
 
15314594
View Documents Promotion Details
Invitrogen™
NW00087BOX
17-well
£141.00
Pack of 10
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
Description
Show More
Specifications

Specifications

Mini Gel Tank, XCell SureLock Mini-Cell
Mini
Bis-Tris Plus
3.5 to 260 kDa
Molecular Weight
10 gels (1 box)
Store at 4–25°C. Do not freeze.
8 cm
8%
1.0 mm
Denaturing
Denaturing
Bolt™, WedgeWell™
Room Temperature
8 cm
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 16 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.