missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bolton-Hunter Reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Bolton-Hunter Reagent is used for iodination of proteins that do not contain tyrosine residues, Bolton-Hunter reagent precursor (3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester) is a hydroxysuccinimide ester used as an intermediate for high radioactive radioiodination of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Description | Powder |
For Use With (Application) | For iodination of proteins that do not contain tyrosine residues |
Molecular Weight (g/mol) | 263.25g/mol, 263.25 |
Product Type | Labeling Reagent |
Quantity | 5 g |
Melting Point | 132° to 133°C |
Content And Storage | −30° to −10°C |
InChI Key | KYRUKRFVOACELK-UHFFFAOYSA-N |
SMILES | OC1=CC=C(CCC(=O)ON2C(=O)CCC2=O)C=C1 |
IUPAC Name | 2,5-dioxopyrrolidin-1-yl 3-(4-hydroxyphenyl)propanoate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'