Learn More
Syngene Bonded Carbon Filter
AirClean™ Systems bonded carbon filters provide for a predictable and reliable gas phase filtration solution.
Brand:  Syngene SYN-ACF43F
Additional Details : Weight : 3.62870kg
 View more versions of this product
Product Code. 17785964
  • £1190.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Description
Description
  • Bonded layered carbon filter for organics and inorganics
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility
Specifications
Specifications
Carbon
Carbon Filter
ACF43F
Silver
Airclean
Activated Carbon
Chemical extraction