Learn More
LTE Scientific™ Boosted Heaters and Pre/Post Vacuum System for the TCR/40/H1 Model
Provides improved air removal at the start of the cycle, ensuring deep steam penetration to eradicate trapped air pockets. Helps speed up the cooling stage when the vacuum is used in conjunction with the boosted heater.
Brand: LTE Scientific™ TCR/40/H5B
Code : NEW
Additional Details : Weight : 260.00000kg
Description
- Vacuum helps speed up cycle times
- Vacuum helps eliminate air pockets within the chamber
- Vacuum assists with the drying cycle
- Boosted heater assists with the drying cycle
Specifications
For Use with TCR/40/H1 | |
TCR/40/H5B | |
Non-sterile |