missing translation for 'onlineSavingsMsg'
Learn More

Borane-d{3}, 1M in tetrahydrofuran

Product Code. 15402997
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15402997

Brand: Thermo Scientific Alfa Aesar 089121.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 13763-62-7 | BD3 | 16.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13763-62-7
Molecular Formula BD3
Molecular Weight (g/mol) 16.85
MDL Number MFCD00064892
InChI Key UORVGPXVDQYIDP-FIBGUPNXSA-N
Synonym borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottles
PubChem CID 11062299
SMILES [2H]B([2H])[2H]

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
UN Number UN3148
Flash Point -21°C (-6°F)
IUPAC Name trideuterioborane
Formula Weight 16.84 as BD3
Concentration or Composition (by Analyte or Components) 1M in tetrahydrofuran
Odor Ether-like
Sensitivity Air & Moisture Sensitive
Density 0.876g/mL
Chemical Name or Material Borane-d_3
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.