missing translation for 'onlineSavingsMsg'
Learn More
Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™ Product Code.: 10685252

Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™

Product Code. 10685252 Shop All Thermo Scientific Chemicals Products
800 mL, AcroSeal™ Glass Bottle, 800mL
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10685252

Brand: Thermo Scientific Acros 185078000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Borane-methyl sulfide complex
Color Colorless to Yellow
Name Note 2M Solution in Tetrahydrofuran
CAS 13292-87-0, 109-99-9, 75-18-3
Molecular Formula C2H9BS
Linear Formula (CH3)2S·BH3
MDL Number MFCD00013189
Concentration 1.80 to 2.20M
Fieser 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Density 0.8500g/mL
Merck Index 15, 1336
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ethyl ether, tetrahydrofurane, hexane,, toluene, methylene chloride and other aprotic, solvents
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
SMILES B.CSC
IUPAC Name (methylsulfanyl)methane borane
Molecular Weight (g/mol) 75.96
PubChem CID 9833925
Formula Weight 75.95
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.85
Quantity 800 mL
Flash Point −17°C
Physical Form Solution
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0472948 Certificate TypeCertificate of Analysis Date25/07/2025 Product Code185078000,185071000
Lot NumberA0471227 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code185078000,185071000
Lot NumberA0469507 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code185078000,185071000
Lot NumberA0463831 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code185078000,185071000
Lot NumberA0463831 Certificate TypeCertificate of Analysis Date09/08/2024 Product Code185078000,185071000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Borane-methyl sulfide complex, 2M solution in tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Borane dimethyl sulfide complex, 2M solution in THF, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.