Learn More
Borane-morpholine complex, 97%, Thermo Scientific Chemicals
£23.32 - £68.54
Chemical Identifiers
CAS | 4856-95-5 |
---|---|
Molecular Formula | C4H12BNO |
Molecular Weight (g/mol) | 100.96 |
MDL Number | MFCD00066736 |
InChI Key | IFVAHVOGOBRFSP-UHFFFAOYSA-N |
Synonym | morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine |
PubChem CID | 6327109 |
IUPAC Name | boron;morpholine |
SMILES | B.C1COCCN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11406840
|
Thermo Scientific Alfa Aesar
L15110.06 |
5 g |
£23.32
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416840
|
Thermo Scientific Alfa Aesar
L15110.14 |
25 g |
£68.54
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4856-95-5 | |
100.96 | |
IFVAHVOGOBRFSP-UHFFFAOYSA-N | |
6327109 | |
B.C1COCCN1 |
C4H12BNO | |
MFCD00066736 | |
morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine | |
boron;morpholine |
Specifications
4856-95-5 | |
Amine-like | |
MFCD00066736 | |
morpholineborane, morpholine borane, borane morpholine complex, borane-morpholine complex, c4h12bno, morpholin-boran, borane morpholine, borane, compound with morpholine, boron; morpholine | |
B.C1COCCN1 | |
100.96 | |
100.96 | |
Borane-morpholine complex |
93°C to 95°C | |
C4H12BNO | |
UN2925 | |
IFVAHVOGOBRFSP-UHFFFAOYSA-N | |
boron;morpholine | |
6327109 | |
97% |
Safety and Handling
GHS H Statement
H228-H301-H314-H318
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302-H312-H314-H335-H500
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 225-450-7
missing translation for 'rtecsNumber' : QD8530000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.