missing translation for 'onlineSavingsMsg'
Learn More

Borane-N,N-diethylaniline complex, 97%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10237697
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 13289-97-9, 91-66-7
Melting Point -30°C to 27°C
Color Colorless to Yellow
Density 0.9200g/mL
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging AcroSeal™ Glass Bottle
Molecular Formula C10H15BN
Linear Formula C6H5N(C2H5)2·BH3
Refractive Index 1.5245 to 1.5265
MDL Number MFCD00013187
Quantity 100 mL
Specific Gravity 0.92
Synonym n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
Solubility Information Solubility in water: may decompose
InChI Key KHYAFFAGZNCWPT-UHFFFAOYSA-N
SMILES [B].CCN(CC)C1=CC=CC=C1
IUPAC Name boron;N,N-diethylaniline
Molecular Weight (g/mol) 160.05
PubChem CID 6335292
Formula Weight 163.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Borane-N, N-diethylaniline complex
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Borane-N,N-diethylaniline complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'