missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%, Thermo Scientific Chemicals

Product Code. 11426790
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridine-borane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 0.92
Boiling Point 65°C (1 mmHg)
Flash Point 73°C (163°F)
Odor Unpleasant
Refractive Index 1.532
Quantity 10 g
UN Number UN2810
Beilstein 3674913
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes slowly in water.
Formula Weight 92.93
Percent Purity 95%
Chemical Name or Material Borane-pyridine complex
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'