Learn More
Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals
£45.70 - £140.00
Chemical Identifiers
CAS | 7337-45-3, 75-64-9 |
---|---|
Molecular Formula | C4H14BN |
Molecular Weight (g/mol) | 86.97 |
MDL Number | MFCD00075635 |
InChI Key | GKFJEDWZQZKYHV-UHFFFAOYSA-N |
Synonym | tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane |
PubChem CID | 6364547 |
IUPAC Name | boron;2-methylpropan-2-amine |
SMILES | B.CC(C)(C)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10694672
|
Thermo Scientific Acros
177300250 |
25 g | Glass Bottle |
£45.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10469650
|
Thermo Scientific Acros
177301000 |
100 g | Glass bottle |
£140.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7337-45-3, 75-64-9 | |
86.97 | |
GKFJEDWZQZKYHV-UHFFFAOYSA-N | |
6364547 | |
B.CC(C)(C)N |
C4H14BN | |
MFCD00075635 | |
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane | |
boron;2-methylpropan-2-amine |
Specifications
7337-45-3, 75-64-9 | |
White | |
95% | |
(CH3)3CNH2·BH3 | |
11,87 | |
Solubility in water: 2.7g/100mL (20°C) | |
B.CC(C)(C)N | |
86.97 | |
86.97 | |
Fine Crystalline Powder |
96°C to 101°C | |
Authentic | |
C4H14BN | |
MFCD00075635 | |
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane | |
GKFJEDWZQZKYHV-UHFFFAOYSA-N | |
boron;2-methylpropan-2-amine | |
6364547 | |
95% | |
Borane-tert-butylamine complex, 95% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable solid.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 230-851-5
missing translation for 'rtecsNumber' : EO3900000
missing translation for 'tsca' : TSCA
- Borane-tert-butylamine complex
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only