missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.0 Product Code.: 15474559

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.0

Product Code. 15474559
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15474559

Brand: Thermo Scientific Chemicals J62125.AK

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 9.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Borate buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY29K541 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberJ62125.AP,J62125.AK
Lot NumberZ25J615 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberJ62125.AP,J62125.AK
Lot NumberM08I514 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ62125.AP,J62125.AK
Lot NumberM08I513 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number
Lot NumberS19I762 Certificate TypeCertificate of Analysis Date28/07/2022 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.0 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.