missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Borate, 0.5M buffer soln., pH 9.5

£49.55 - £74.17

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444579
View Documents
Thermo Scientific Chemicals
J62154.AK
250 mL
£49.55
250mL
Please to purchase this item. Need a web account? Register with us today!
15454579
View Documents
Thermo Scientific Chemicals
J62154.AP
500 mL
£74.17
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

Solubility
Soluble in water.

Notes
Store at 25°C. This product is relatively stable under this condition. Best result obtained with use within 6-8 months of purchase.
Specifications

Chemical Identifiers

1330-43-4
201.21
NXSVMNYRVBQCMO-UHFFFAOYSA-N
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
B4Na2O7
MFCD00081185
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)

Specifications

1330-43-4
9.5
B4Na2O7
Soluble in water.
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
201.21
0.5 M
Liquid
MFCD00081185
NXSVMNYRVBQCMO-UHFFFAOYSA-N
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Borate buffer soln.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c

H319-H360FD

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only