missing translation for 'onlineSavingsMsg'
Learn More
Borax Anhydrous MP Biomedicals Product Code.: 11491831

Borax Anhydrous MP Biomedicals

Product Code. 11491831 Shop All MP Biomedicals Products
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 11491831

Brand: MP Biomedicals 0219030980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Sodium Tetraborate, Sodium Borate Anhydrous, Sodium Borate
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Quantity 100 g
Formula Weight 201.2
Density 2.367 g/mL
Chemical Name or Material Borax, Anhydrous
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : boric acid disodiumsalt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Borax Anhydrous MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.