missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borer Chemie™ borer 21 LAB CLF
Click to view available options
Quantity:
1 x 6 kg
1 x 12 kg
1 x 32 kg
Unit Size:
12kg
32kg
6kg
Description
- Concentrate for automated cleaning
- High yield, typical concentration: 5-10 mL/L
- Excellent material compatibility, also for aluminium
- Broad cleaning spectrum
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Without chlorine, EDTA, NTA and surfactants
- Acid rinse with borer cleaner recommended
- Technical support by the manufacturer
Specifications
Specifications
| Form | Liquid |
| pH | ∼11.4 (1% Solution) |
| Composition | Alkalis, complexing agents, corrosion inhibitor, sequestering agents |
| Concentration | Concentrate |
| Container Type | Canister |
| Content And Storage | 5°C to 25°C |
| Quantity | 1 x 6 kg |
| Volume (English) | 1.2 gal. |
| Volume (Metric) | 4.4 L |
| Weight (English) | 13.2 lb. |
| Show More |
Safety and Handling
Product Identifier
- 6KG borer 21 LAB CLF
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
- MIXTURE LIST-Contains : TETRAPOTASSIUM PYROPHOSPHATE, DIPOTASSIUM TRIOXOSILICATE, SODIUM HYPOCHLORITE
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction