missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ borer 25 ORGANACID

Cleaner and neutralizer for laboratory washers. Shortens rinsing phases and reduces water consumption. borer 25 ORGANACID is phosphate-free and suitable for environmental and water laboratories. Pre-rinsing agent against acid-soluble residues.

Brand:  Borer Chemie™ 502100.15-K10R

 View more versions of this product

Product Code. 17415112

  • £125.00 / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Neutralizer and cleaner for automated use
  • Ecological solution without phosphates and surfactants
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Typical application concentration: 3 mL/L
  • Excellent material compatibility, also for aluminium
  • Can be rinsed off without leaving residues
  • Also as pre-rinsing agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications

Specifications

Liquid
Organic acid
Canister
1 x 10 kg
8.5 L
10 kg
∼2.6 (1% Solution)
Concentrate
5°C to 25°C
2.2 gal.
22 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.