missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Lewis acid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 3.6 to 4.0 (4% at 20°C) |
Color | White |
Melting Point | 169.0°C |
Heavy Metals (as Pb) | 5ppm max. |
Non_volatile Matter | 0.05% max. |
Packaging | Plastic bottle |
Assay Percent Range | 99.5% |
Quantity | 2.5 kg |
Linear Formula | H3BO3 |
Physical Form | Fine Crystals |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'