missing translation for 'onlineSavingsMsg'
Learn More
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals Product Code.: 10307121

Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Product Code. 10307121
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 10307121

Brand: Thermo Scientific Acros 327130010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer, Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color White
Melting Point 169.0°C
Heavy Metals (as Pb) 50ppm max.
Assay Percent Range 99.5%
Quantity 1 kg
Linear Formula H3BO3
Physical Form Powder
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Solubility Information Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
Formula Weight 61.83
Percent Purity 99.5%
Grade Molecular Biology
Chemical Name or Material Boric acid, DNAse, RNAse and Protease Free
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458828 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number327132500,327130010
Lot NumberA0461503 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number327132500,327130010
Lot NumberA0458828 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number327132500,327130010
Lot NumberA0457107 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number327132500,327130010
Lot NumberA0443856 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number327132500,327130010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric acid, 99.5%, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.