Learn More
Boric acid, 99+%, Thermo Scientific Chemicals
Description
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Boric acid is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is employed in leather manufacture, electroplating and cosmetics. It is involved in the production of monofilament fiberglass which finds applications in boats, industrial piping, LCD flat panel displays and computer circuit boards. It is in combination with sodium tetraborate decahydrate (borax) and is used as a welding flux by blacksmiths. The mixture of boric acid and silicone oil is useful in the production of silly putty.
Solubility
Soluble in water and alcohols. Slightly soluble in acetone and pyridine.
Notes
Store in cool place. Moisture sensitive. Incompatible with potassium and acid anhydrides.

Specifications
Specifications
pH | 3.8 to 4.8 |
Melting Point | ∼185°C (decomposition) |
Density | 1.435 |
Odor | Odorless |
Flash Point | None |
Quantity | 2500 g |
Linear Formula | H3BO3 |
Beilstein | 1697939 |
Merck Index | 14,1336 |
Solubility Information | Soluble in water and alcohols. Slightly soluble in acetone and pyridine. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z31K005 | Certificate of Analysis | 27/01/2025 | A10896.0C,A10896.36,A10896.0E |
10251263 | Certificate of Analysis | 30/08/2024 | A10896.0C,A10896.36,A10896.0E |
10252057 | Certificate of Analysis | 14/08/2024 | A10896.0C,A10896.36,A10896.0E |
10249074 | Certificate of Analysis | 19/12/2023 | A10896.0C,A10896.36,A10896.0E |
S09J032 | Certificate of Analysis | 26/07/2023 | A10896.0C,A10896.36,A10896.0E |
Safety and Handling

- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Boric acid (H3BO3)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.