missing translation for 'onlineSavingsMsg'
Learn More
Boric Acid, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10682251

Boric Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. 10682251
500 g
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
This item is not returnable. View return policy

Product Code. 10682251

Brand: Fisher Chemical B/3800/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

H3BO3, CAS Number-10043-35-3, 500g, White, 61.83g/mol, 169 deg.C, 11337, Plastic powder jar, 3.8 to 4.8, Powder Solid, 2.7mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.8 to 4.8
Color White
Melting Point 169°C
Packaging Plastic powder jar
Vapor Pressure 2.7mbar at 20°C
Quantity 500 g
Physical Form Powder Solid
Formula Weight 61.83g/mol
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : boric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric Acid, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.