missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
This high-purity electrophoresis-grade reagent is used in the preparation of Tris/Borate/EDTA electrophoresis buffers.

Specifications
Specifications
pH | 3.8 to 4.8 |
Color | White |
Melting Point | 169°C |
Heavy Metals (as Pb) | 0.001% max. |
Identification | Pass Test |
Non_volatile Matter | 0.05% max. |
Packaging | Glass Bottle |
Vapor Pressure | 2.7mbar at 20°C |
Assay Percent Range | ≥99 % |
Quantity | 500 g |
Show More |
Safety and Handling

Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction