missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Granular, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
£121.00 - £416.00
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15568214
|
Avantor J.T.Baker
|
500 g | Poly Bottle |
£121.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578214
|
Avantor J.T.Baker
|
2.5 kg | Poly Bottle |
£416.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Molecular Biology Buffers
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
10043-35-3 | |
BH3O3 | |
MFCD00011337 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Boric Acid |
1L = 1.43kg | |
Granular | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
Ultrapure Bioreagent |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Boric Acid
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Boric acid