Learn More
Boric Acid, Powder 99+%, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
£49.07 - £281.40
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103820
|
Thermo Scientific Acros
180570010 |
1 kg | Plastic bottle |
£49.07
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10052390
|
Thermo Scientific Acros
180570025 |
2.5 kg | Plastic bottle |
£99.77
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904101
|
Thermo Scientific Acros
180570100 |
10 kg | Plastic drum |
£281.40
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
White | |
169.0°C | |
99+% | |
H3BO3 | |
MFCD00011337 | |
15, 1339 | |
Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol. | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
≥99% | |
Boric acid |
10043-35-3 | |
Odorless | |
BH3O3 | |
Coarse Crystals | |
01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83 | |
Extra Pure |
Safety and Handling
GHS P Statement May damage fertility. May damage the unborn child.
Danger
- Boric acid
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.