missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 160°C (decomposition)(literature) |
CAS Max % | 100.0000% |
Density | 1.510 g/cm3 |
Packaging | Paper / Poly Bag |
Vapor Pressure | 2.6mmHg (20°C) |
Assay Percent Range | 99.5 to 100.5% |
Quantity | 25 kg |
Linear Formula | H3BO3 |
Physical Form | powder |
UN Number | NONH for all modes of transport |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'