missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%, Honeywell Fluka™

Catalog Number 15683280
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
This item is not returnable. View return policy
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Vapor Pressure 2.6mmHg (20°C)
Quantity 6 x 2.5 kg
Linear Formula H3BO3
UN Number NONH for all modes of transport
Formula Weight 61.83
Percent Purity ≥99.8%
Grade ACS/puriss. p.a.
Chemical Name or Material Boric Acid, Reag. Ph. Eur. reag. ISO
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'