missing translation for 'onlineSavingsMsg'
Learn More

Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%, Honeywell Fluka™

Product Code. 15653280
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance,≥99.8%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Melting Point 160°C (decomposition)(literature)
CAS Max % 100.0000%
Density 1.510 g/cm3
Packaging Fiberboard Box
Vapor Pressure 2.6mmHg (20°C)
Quantity 25 kg
Linear Formula H3BO3
UN Number NONH for all modes of transport
Formula Weight 61.83
Percent Purity ≥99.8%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Chemical Name or Material Boric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., buffer substance, ≥99.8%
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.