missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Reagent Grade, ≥99.5%, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
10 kg
Packaging:
Drum with inliner
Poly Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
pH | 3.6 to 4 (at 20°C) |
Color | Colorless |
Melting Point | 160°C |
Density | 1.510g/cm3 at 20°C |
Odor | Odorless |
Packaging | Drum with inliner |
Vapor Pressure | 2.6mmHg (20°C) |
Quantity | 10 kg |
Linear Formula | H3BO3 |
Physical Form | Solid |
Show More |
Safety and Handling
Product Identifier
- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'