missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, Reagent Grade, ≥99.5%, Honeywell™

Product Code. 15687680
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
10 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

pH 3.6 to 4 (at 20°C)
Color Colorless
Melting Point 160°C
Density 1.510g/cm3 at 20°C
Odor Odorless
Packaging Poly Bottle
Vapor Pressure 2.6mmHg (20°C)
Quantity 500 g
Linear Formula H3BO3
Physical Form Solid
UN Number NONH for all modes of transport
Formula Weight 61.83g/mol
Percent Purity ≥99.5%
Chemical Name or Material Boric acid
Show More Show Less
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'