missing translation for 'onlineSavingsMsg'
Learn More

(-)-Borneol, 97+%

Product Code. 11400583
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11400583

Brand: Thermo Scientific Alfa Aesar A12684.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 464-45-9 | C10H18O | 154.25 g/mol

(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Borneol is used to prepare its esters by reacting with acids. Its derivatives are used as chiral ligands in asymmetric synthesis. It is also used in flavors and perfumes. Further, it is used in traditional Chinese medicine as moxa. In addition to this, it is used as a component of many essential oils and also used as a natural insect repellent.

Solubility
Soluble in chloroform, ethanol, acetone, ether, benzene, toluene, decalin and tetralin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-45-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00003759,MFCD00066426,MFCD00066427
InChI Key DTGKSKDOIYIVQL-NQMVMOMDSA-N
Synonym --borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
PubChem CID 1201518
ChEBI CHEBI:15394
SMILES CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Melting Point 208°C to 211°C
Boiling Point 210°C
Flash Point 65°C (149°F)
Assay Percent Range 97+%
Quantity 50 g
UN Number UN1312
Beilstein 3587558
Merck Index 14,1338
Solubility Information Soluble in chloroform,ethanol,acetone,ether,benzene,toluene,decalin and tetralin. Insoluble in water.
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
Formula Weight 154.25
Percent Purity ≥97%
Chemical Name or Material (-)-Borneol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (-)-Borneol
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (-)-Borneol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.