missing translation for 'onlineSavingsMsg'
Learn More

Boron carbide, 90+%, Thermo Scientific Chemicals

£36.28 - £137.69

Chemical Identifiers

CAS 12069-32-8
Molecular Formula CB4
Molecular Weight (g/mol) 55.25
MDL Number MFCD00011520
InChI Key INAHAJYZKVIDIZ-UHFFFAOYSA-N
Synonym boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
PubChem CID 123279
IUPAC Name 2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
SMILES B12B3B4B1C234
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361448
View Documents
Thermo Scientific Alfa Aesar
010922.22
100 g
£36.28
100g
Please to purchase this item. Need a web account? Register with us today!
11371448
View Documents
Thermo Scientific Alfa Aesar
010922.36
500 g
£137.69
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12069-32-8
55.25
INAHAJYZKVIDIZ-UHFFFAOYSA-N
123279
B12B3B4B1C234
CB4
MFCD00011520
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane

Specifications

2350°C
Black
>90%
MFCD00011520
Insoluble in water.
B12B3B4B1C234
55.25
55.26
2.52
12069-32-8
Powder
CB4
boron carbide, boron carbide b4c, tetra 10b boron carbide, boron carbide, dust, tetrabor, boron carbide powder, boron carbide b4c grade hp, boron carbide b4c grade hs, b4-c, boron carbide b4c grade hd 07
INAHAJYZKVIDIZ-UHFFFAOYSA-N
2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
123279
Odorless
Boron carbide
Videos
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H332-H335

missing translation for 'einecsNumber' : 235-111-5

missing translation for 'rtecsNumber' : ED7420000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only