missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, 98%, Thermo Scientific Chemicals
£110.55 - £397.98
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
SMILES | B=N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15580544
|
Thermo Scientific Acros
441800500 |
50 g |
£110.55
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15590544
|
Thermo Scientific Acros
441802500 |
250 g |
£397.98
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 |
Specifications
2700.0°C to 3000.0°C | |
97.5% min | |
BN | |
15,1347 | |
Solubility in water: slightly soluble (20°C). Other solubilities: soluble in water (95°) | |
B=N | |
24.82 | |
CHEBI:50883 | |
98% |
10043-11-5 | |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
boranylidyneamine | |
66227 | |
24.82 | |
Boron nitride |
Safety and Handling
missing translation for 'einecsNumber' : 233-136-6
Product Identifier
- Boron nitride
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boron nitride, 98%, Thermo Scientific Chemicals