Learn More
Boron nitride, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Boron nitride used in the manufacture of high-temperature equipment parts due to its excellent thermal and chemical stability. It is used as a lubricant and an additive to cosmetics, paints, dental cements and pencil leads. It is also used to provide self lubricating properties to ceramics, alloys, resins, plastics and rubbers. In addition to this, it has many industrial applications such as xerographic process, laser printers, oxygen sensors and proton conductors. Cubic boron nitride is used as an abrasive material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.5% (metals basis) |
Solubility Information | Insoluble in water. |
IUPAC Name | azanylidyneborane |
Formula Weight | 24.82 |
Chemical Name or Material | Boron nitride |
RUO – Research Use Only